Gelex secondary standards kit for 2100 series turbidity meters
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2464105 2589200 2589000 2620000
HACH2464105EA 396 EUR
HACH2464105 705-0974 705-0973 HACH2620000
Gelex secondary standards kit for 2100 series turbidity meters
Standaarden Standaarden voor troebelheidsbepaling
Gelex secondary standards kit for 2100P and 2100Q series portable turbidimeters (includes 1 sealed vial each for 0 to 10 NTU, 10 to 100 NTU, 100 to 1000 NTU).
Gelex secondary standards kit for 2100N and 2100N IS series laboratory turbidimeters (includes 1 sealed vial each, Stray Light Standard, 0 to 2 NTU, 0 to 20 NTU, 0 to 200 NTU, 200 to 4000 NTU).
Gelex secondary standards kit for for 2100AN and 2100AN IS laboratory turbidimeter (includes 1 sealed vial each, Stray Light Standard, 0 to 2 NTU, 0 to 20 NTU, 0 to 200 NTU, 200 to 4000 NTU, 4000 to 10000 NTU).