MXP high pressure switching valves, Rheodyne®, MX Series II™
Leverancier: IDEX RHEODYNE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MXP9900-000 MXP7900-000
554-2669EA 3280 EUR
554-2669 554-2666
MXP high pressure switching valves, Rheodyne®, MX Series II™
Ventielen Regelventielen
These high pressure actuated valves are designed to be added to an existing instrument or to work optimally with stand-alone lab configurations.
  • Can withstand up to 6000 psi (414 bar)
  • Chemically compatible for use with most mobile phase compositions
  • Available with analytical and nano-scale flow paths
  • Multiple control options
  • Stand alone capability
  • Included software