Shirts, long sleeves, Trapani 2673
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2673/BS0/1XL 2673/BS0/2XL 2673/BS0/3XL 2673/BS0/L 2673/BS0/M 2673/BS0/S
SIOE2673/BS0/1XLEA 36.3 EUR
SIOE2673/BS0/1XL SIOE2673/BS0/2XL SIOE2673/BS0/3XL SIOE2673/BS0/L SIOE2673/BS0/M SIOE2673/BS0/S
Shirts, long sleeves, Trapani 2673
Werkkleding
Longsleeve made of 50% Viloft® and 50% polyester (interlock). The fabric features an excellent thermal performance and has high moisture wicking properties. It therefore keeps the body warm and dry.
  • Long sleeves
  • Ribbed collar, round neck
  • Seamless
  • Improved comfort due to wristband
  • Stitched seams
Weight: 210 g/m²