High visibility overall, class 3, 8601 TH
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1000041712XL 1000041713XL 100004171L 100004171M 100004171S 100004171XL
KWIN1000041712XLEA 138.9 EUR
KWIN1000041712XL KWIN1000041713XL KWIN100004171L KWIN100004171M KWIN100004171S KWIN100004171XL
High visibility overall, class 3, 8601 TH
Werkkleding
High visibility overalls made of 70% polyester and 30% cotton.
  • Front placket opening with two-way zip to top of collar and snap fastening
  • Two chest pockets with flap and velcro fastener, two front pockets, back pockets, inside pocket with button
  • Adjustable cuffs, adjustable elasticated back
  • D-ring, side openings
  • Leg pocket with flap and velcro fastening, phone pocket with flap and velcro fastening
  • Folding rule pocket with pen pocket, and button and loop for sheath knife
  • Knee pockets with opening for height adjustable knee pads
  • Reflective tape on shoulders
Weight: 300 g/m²
Certificaten: EN ISO 20471 (class 3)