Rain coats, Gascogne 3729
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3792/B90/1XL 3792/B90/2XL 3792/B90/3XL 3792/B90/L 3792/B90/M 3792/B90/S
SIOE3792/B90/1XLEA 86.9 EUR
SIOE3792/B90/1XL SIOE3792/B90/2XL SIOE3792/B90/3XL SIOE3792/B90/L SIOE3792/B90/M SIOE3792/B90/S
Rain coats, Gascogne 3729
Werkkleding
Wind- and waterproof coat made of Flexothane® Classic, a polyamide knitting with PU coating. The fabric is lightweight, noiseless and provides extended durability.
  • High frequency welded seams
  • Straight collar with foldaway hood in collar
  • Closure with press studs
  • Two patched pockets
  • Raglan sleeves with elasticated wind cuffs
Back length: 120 cm
Weight: 180 g/m²
Certificaten: ENV 343 (3-2)