Bodywarmer, Classic, S415
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S415NARXL S415NARL S415NARM S415NARS S415NARXXL S415NARXXXL S415BKRXS S415BKRM S415BKRS S415BKRL S415BKRXL S415BKRXXL S415BKRXXXL
PRTWS415NARXLEA 38.3 EUR
PRTWS415NARXL PRTWS415NARL PRTWS415NARM PRTWS415NARS PRTWS415NARXXL PRTWS415NARXXXL PRTWS415BKRXS PRTWS415BKRM PRTWS415BKRS PRTWS415BKRL PRTWS415BKRXL PRTWS415BKRXXL PRTWS415BKRXXXL
Bodywarmer, Classic, S415
Werkkleding
This classic style bodywarmer is made of 100% PVC coated polyester. The polyester lining and wadding provide an extra layer of warmth. Ideal for outdoor applications.
  • Zip front fastening with studded storm flap
  • Left chest pen pocket with mobile phone pocket
  • Two lower patch pockets with Velcro fastened flaps, two side hand warmer pockets
  • Elasticated drawcord hem with spring loaded toggles
  • Generously cut for a relaxed fit over workwear
  • Showerproof
Weight (outer fabric): 160 g/m²
Weight (polyester lining): 60 g/m²
Weight (polyester wadding): 200 g/m²