High visibility softshell jacket, two tone, S429
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S429YERL S429YERM S429YERS S429YERXL S429YERXXL S429YERXXXL
PRTWS429YERLEA 90 EUR
PRTWS429YERL PRTWS429YERM PRTWS429YERS PRTWS429YERXL PRTWS429YERXXL PRTWS429YERXXXL
High visibility softshell jacket, two tone, S429
Werkkleding
This softshell jacket offers comfort and a fresh new look. The polyester/spandex fabric bonded with 100% micro polar fleece is lightweight but gives added warmth during cold weather.
  • Two waterproof side zip pockets
  • Reflective tape (5 cm)
  • Velcro adjustable cuffs
  • Contrast panels
Weight: 310 g/m²
Certificaten: EN ISO 40271 (Class 3)