Material pouch, snikki
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100014240 100015940
KWIN100014240EA 46.9 EUR
KWIN100014240 KWIN100015940
Material pouch, snikki
Zakken Pouches and Belt Cases
Pouch are made of heavy duty split leather that consist of two pockets for screws and nails with sheath knife holder, rivet reinforced and easy quick-grip.
Big pocket: 160×80×200 mm (W×D×H)
Small pocket: 140×45×130 mm (W×D×H)