Flame-retardant high visibility jacket, BizFlame™ FR61
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR61YNRL FR61YNRM FR61YNRS FR61YNRXL FR61YNRXXL FR61YNRXXXL
PRTWFR61YNRLEA 181 EUR
PRTWFR61YNRL PRTWFR61YNRM PRTWFR61YNRS PRTWFR61YNRXL PRTWFR61YNRXXL PRTWFR61YNRXXXL
Flame-retardant high visibility jacket, BizFlame™ FR61
Werkkleding
This multi-norm jacket is made of flame-retardant, anti-static BizFlame™ fabric (99% polyester, 1% carbon fibre).
  • Flame-resistant reflective striping for improved visibility is double stitched
  • Triple stitched seams
  • Concealed 2-way front brass zip
  • Two concealed front chest pockets with piping detail, radio loop
The fabric provides protection against electric arcs and open flames. Also suitable for welding applications.
Weight: 345 g/m²
Certificaten: EN ISO 20471 (Class 3), EN 1149-5, EN 13034 (type 6), EN ISO 11612 (A1 + A2, B1, C1, E3, F1), ENMore Product Information