Overalls with bib and braces, Toscana 4644
Leverancier: Sioen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIOE4644/140/1XLEA 78.1 EUR
SIOE4644/140/1XL SIOE4644/140/2XL SIOE4644/140/S SIOE4644/140/3XL SIOE4644/140/L SIOE4644/140/M
Overalls with bib and braces, Toscana 4644
Werkkleding
Rain protective trousers made of two different fabrics: The upper part is made of Flexothane® 2000 (100% polyamide knitting with PU coating), the lower part is made of SIONYL (100% polyester fabric with PVC coating).
  • Elastic braces with PVC buttons and buckles
  • Reinforced legs, knees reinforced with foam padding
  • Front bib
  • High frequency welded seams
Weight Flexothane® 2000: 200 g/m²
Weight SIONYL: 420 g/m²
Certificaten: EN 343 (3-1)
Oeko-Tex® Standard 100