Shut-off valves, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P-782 P-782NF P-783 P-783-01
554-3193EA 128 EUR
554-3193 554-3194 554-3195 554-3196
Shut-off valves, Upchurch Scientific®
Ventielen Terugslagventielen
Stop a flow stream quickly with these shut-off valves. The bodies are manufactured from either PEEK or ETFE. Both versions feature a PCTFE rotor, making them highly resistant to chemicals. The blue colorant used in some valve configurations has proven not to leach out with common HPLC solvents.
  • For low and medium pressure applications
  • Biocompatible, all-polymer flow path
  • Pressure rated to 500 psi (34 bar)