Overalls, Herford 6218
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6218/A41/1XL 6218/A41/2XL 6218/A41/3XL 6218/A41/L 6218/A41/M 6218/A41/S 6218/B90/1XL 6218/B90/2XL 6218/B90/3XL 6218/B90/L 6218/B90/M 6218/B90/S
SIOE6218/A41/1XLEA 86.9 EUR
SIOE6218/A41/1XL SIOE6218/A41/2XL SIOE6218/A41/3XL SIOE6218/A41/L SIOE6218/A41/M SIOE6218/A41/S SIOE6218/B90/1XL SIOE6218/B90/2XL SIOE6218/B90/3XL SIOE6218/B90/L SIOE6218/B90/M SIOE6218/B90/S
Overalls, Herford 6218
Werkkleding
Lightweight, supple overalls made of Flexothane® Essential fabric consisting of 100% polyester knitting with PU coating. The fabric is waterproof and windproof, and features high frequency welded seams.
  • Fixed hood, zip closure under double flap with press studs
  • Raglan sleeves
  • Sleeve and ankle narrowing by press studs
Weight: 170 g/m²
Certificaten: ENV 343 (3-1)