VWR® bolpipetten, twee merken, klasse AS
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
612-4775EA 21.4 EUR
612-4775 612-4776 612-4777 612-4778 612-4779 612-4780 612-4781 612-4782 612-4783 612-4784
VWR® bolpipetten, twee merken, klasse AS
Pipetten Volpipetten
Borosilicaat 9.1 glas.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in high contrast amber stain
Certificaten: DIN EN ISO 648
Bestelinformatie: Dated batch identification and batch certificate.