Crystallisation plates, CrystalStar™, 60- and 72-well Terasaki plates
Leverancier: Greiner Bio-One
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
653102_580 654102_270
737-3526EA 608 EUR
737-3526 737-3527
Crystallisation plates, CrystalStar™, 60- and 72-well Terasaki plates
Microplaten Kristalllisatieplaten
Terasaki plates are widely used for microbatch crystallisation.
  • Crystallisation drop is localised centrally as a result of the conical well geometry, and the flat well bottom makes for optimal monitoring
  • The rim of the plate makes it possible to fill all of the wells with oil at the same time
  • Supplied with fitting lid