Rain trousers, Tomar 654Z, S.E.P.P.
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
654Z/NS0/XS 654Z/NS0/S 654Z/NS0/M 654Z/NS0/L 654Z/NS0/1XL 654Z/NS0/2XL 654Z/NS0/3XL
113-07421EA 83.5 EUR
113-07421 113-07422 113-07423 113-07424 113-07425 113-07426 113-07427
Rain trousers, Tomar 654Z, S.E.P.P.
Werkkleding
Highly breathable, water- and windproof trousers made of Siopor® Extra consisting of 100% polyester fabric (cotton touch) laminated with 100% PU film. The fabric is supple and lightweight. With taped seams and fixed polyamide lining.
  • Two inset pockets, one patched pocket on the thigh
  • Ankles adjustable by touch and close fastening
  • Elasticated waist and drawstring
Weight: 205 g/m²
Certificaten: EN 343 (3-3)