High visibility rain trousers, Batur 6564
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6564/FC1/1XL 6564/FC1/2XL 6564/FC1/3XL 6564/FC1/L 6564/FC1/M 6564/FC1/S
SIOE6564/FC1/1XLEA 49.9 EUR
SIOE6564/FC1/1XL SIOE6564/FC1/2XL SIOE6564/FC1/3XL SIOE6564/FC1/L SIOE6564/FC1/M SIOE6564/FC1/S
High visibility rain trousers, Batur 6564
Werkkleding
Water- and windproof trousers made of stretchable Flexothane® Essential consisting of 100% polyester knitting with PU coating. With high frequency welded seams.
  • Provide good cold protection
  • Legs adjustable by press studs
  • Elasticated waist
Weight: 170 g/m²
Certificaten: EN 471 (1-2), ENV 343 (3-1)