Rain trousers, Surakarta 6604
Leverancier: Sioen
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Flexothane® Essential
SIOE6604/Y58/LEA 29.9 EUR
SIOE6604/Y58/L SIOE6604/Y58/M SIOE6604/A41/1XL SIOE6604/A41/S SIOE6604/A41/3XL SIOE6604/A41/2XL SIOE6604/A41/M SIOE6604/Y58/S SIOE6604/A41/L SIOE6604/Y58/2XL SIOE6604/Y58/3XL SIOE6604/Y58/1XL
Rain trousers, Surakarta 6604
Werkkleding
Water- and windproof rain trousers made of Flexothane® Essential (100% polyester knitting with PU coating).
  • High frequency welded seams
  • Front fly with press studs, two side slits
  • Legs adjustable by press studs
  • Drawstring in waist
Weight: 170 g/m²
Certificaten: ENV 343 (3-1)