Laboratory coats, Standard, C852
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C852WHRL C852WHRM C852WHRS C852WHRXL C852WHRXXL C852WHRXXXL C852NARL C852NARXL C852NARM C852NARS C852NARXXL C852NARXXXL C852NAR4XL
PRTWC852WHRLEA 33.8 EUR
PRTWC852WHRL PRTWC852WHRM PRTWC852WHRS PRTWC852WHRXL PRTWC852WHRXXL PRTWC852WHRXXXL PRTWC852NARL PRTWC852NARXL PRTWC852NARM PRTWC852NARS PRTWC852NARXXL PRTWC852NARXXXL PRTWC852NAR4XL
Laboratory coats, Standard, C852
Werkkleding
Standard coats made of Kingsmill fabric (65% polyester and 35% cotton). The fabric is pre-shrunk and has superior dye retention.
  • Chest pocket, two lower patch pockets
  • Concealed stud front closure
  • Back vent
Weight: 245 g/m²