High visibility rain overalls, Etna 6936
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6936/FC1/1XL 6936/FC1/2XL 6936/FC1/3XL 6936/FC1/L 6936/FC1/M 6936/FC1/S 6936/FY1/1XL 6936/FY1/2XL 6936/FY1/3XL 6936/FY1/L 6936/FY1/M 6936/FY1/S
SIOE6936/FC1/1XLEA 140 EUR
SIOE6936/FC1/1XL SIOE6936/FC1/2XL SIOE6936/FC1/3XL SIOE6936/FC1/L SIOE6936/FC1/M SIOE6936/FC1/S SIOE6936/FY1/1XL SIOE6936/FY1/2XL SIOE6936/FY1/3XL SIOE6936/FY1/L SIOE6936/FY1/M SIOE6936/FY1/S
High visibility rain overalls, Etna 6936
Werkkleding
Water- and windproof overalls made of Flexothane Classic, 100% polyamide knitting with PU coating.
  • High frequency welded seams
  • Fixed hood, zip closure under flap with press studs
  • Legs adjustable by press studs
  • Elasticated waist
The lightweight garment features an extended durability and provides protection against liquid chemicals.
Weight: 180 g/m²
Certificaten: EN 14786, EN 14605 (type 4), EN 14126 (type 4-B), EN 20471 (Class 3), EN 343 (41X))