Chemical resistant overalls, type 5/6, BizTex® SMS, ST30
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST30NARXL ST30NARL ST30NARM ST30NARXXL ST30WHRL ST30WHRM ST30WHRS ST30WHRXL ST30WHRXXL ST30WHRXXXL ST30NARXXXL
PRTWST30NARXLEA 215 EUR
PRTWST30NARXL PRTWST30NARL PRTWST30NARM PRTWST30NARXXL PRTWST30WHRL PRTWST30WHRM PRTWST30WHRS PRTWST30WHRXL PRTWST30WHRXXL PRTWST30WHRXXXL PRTWST30NARXXXL
Chemical resistant overalls, type 5/6, BizTex® SMS, ST30
Wegwerpkleding
These generous overalls made of SMS fabric fit easily over work clothes and offer excellent protection to type 5/6 hazards.
  • Durable, high tear and abrasion resistance
  • Breathable, anti-static
  • Elasticated hood, waist, wrist and ankles
Weight: 55 g/m²
Certificaten: EN 1149-5, EN 1073-2, EN 13034 (type 6), EN ISO 13982 (type 5)