Chemical resistant overalls, BizTex® Microporous, ST40
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST40WHRL ST40WHRM ST40WHRXL ST40WHRXXL ST40WHRS ST40WHRXXXL
PRTWST40WHRLEA 214 EUR
PRTWST40WHRL PRTWST40WHRM PRTWST40WHRXL PRTWST40WHRXXL PRTWST40WHRS PRTWST40WHRXXXL
Chemical resistant overalls, BizTex® Microporous, ST40
Wegwerpkleding
This overall made of BizTex® Microporous fabric (PP/PE) offers excellent protection against airborne particulates and liquid chemicals.
  • Elasticated hood, legs, cuffs and waist
  • Two way front zip fastening with sealable front flap
  • Spunbonded PE fibres provide toughness and and an excellent barrier
Weight: 60 g/m²
Certificaten: EN 13034 (Type 6), EN ISO 13982 (Type 5), EN 1149-5, EN 14126, EN 1073-2, Cat. III