Chemical resistant overalls, BizTex® Microporous, ST60
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ST60WHRL ST60WHRM ST60WHRXL ST60WHRXXL
PRTWST60WHRLEA 513 EUR
PRTWST60WHRL PRTWST60WHRM PRTWST60WHRXL PRTWST60WHRXXL
Chemical resistant overalls, BizTex® Microporous, ST60
Wegwerpkleding
This overall made of BizTex® Microporous fabric offers excellent chemical and dust protection from biological agents.
  • Ultra low linting with bound tape seams: extremely effective barrier to chemicals
  • Ergonomically design for unrestricted movement comfort and breathability
  • Elasticated 3-piece hood with chinflap
Weight: 60 g/m²
Certificaten: EN 14605 (Type 4), EN 13034 (type 6), EN ISO 13982 (type 5), EN 1149-5, EN 1073-2, EN 14126, Cat. III