High visibility executive waistcoat, S476
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S476ORRL S476ORRM S476ORRS S476ORRXL S476ORRXXL S476ORRXXXL S476YERL S476YERM S476YERS S476YERXL S476YERXXL S476YERXXXL S476YER4XL S476YER6XL S476YER5XL S476YER7XL S476YERXS
PRTWS476ORRLEA 15.5 EUR
PRTWS476ORRL PRTWS476ORRM PRTWS476ORRS PRTWS476ORRXL PRTWS476ORRXXL PRTWS476ORRXXXL PRTWS476YERL PRTWS476YERM PRTWS476YERS PRTWS476YERXL PRTWS476YERXXL PRTWS476YERXXXL PRTWS476YER4XL PRTWS476YER6XL PRTWS476YER5XL PRTWS476YER7XL PRTWS476YERXS
High visibility executive waistcoat, S476
Werkkleding
This executive vest made of 100% polyester combines the light weight of a waistcoat with the practicality of pockets for those situations when a jacket may be too warm.
  • Two side pockets with flaps, mobile phone and ID pocket
  • HiVisTex reflective tape (5 cm)
  • Front zip fastening
Certificaten: EN ISO 20471 (Class 2); GO/RT 3279 Issue 8 (orange only)