High visibility two tone jacket, 4-in-1, S769
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S769YNRXXL S769YNRXL S769YNRL S769YNRM S769ONRXXL S769ONRXL S769ONRS S769ONRM S769ONRL S769YNRS
PRTWS769YNRXXLEA 89.7 EUR
PRTWS769YNRXXL PRTWS769YNRXL PRTWS769YNRL PRTWS769YNRM PRTWS769ONRXXL PRTWS769ONRXL PRTWS769ONRS PRTWS769ONRM PRTWS769ONRL PRTWS769YNRS
High visibility two tone jacket, 4-in-1, S769
Werkkleding
Modern, breathable jacket made of 300D Oxford weave polyester. The PU coating makes the fabric durable and waterproof. The water- and stain-repellent coating on the outside helps to keep the garment clean.
  • Detachable, quilted sleeves turn the garment into a body warmer when removed
  • Taped waterproof seams
  • Reversible
  • Zip and stud front fastening
  • Radio pocket, concealed ID holder, mobile phone pocket
Designed to keep you warm and dry, this item provides maximum visibility and is suitable for all weather conditions. With polyester Peach finish in body and polyester lining and wadding in sleeves.
Weight: 190 g/m² (without lining)
Certificaten: EN 471 (1-2)