Rain jackets, 3-in-1, Orkney, S532
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S532GRRXL S532GRR4XL S532GRRXXL S532GRRL S532GRRS S532NARXXXL S532GRRXXXL S532NAR4XL S532NARL S532NARM S532NARS S532NARXL S532NARXXL S532GRRM S532RERL S532RERXL S532RERS S532RERXXL S532RERM S532RERXXXL
PRTWS532GRRXLEA 109 EUR
PRTWS532GRRXL PRTWS532GRR4XL PRTWS532GRRXXL PRTWS532GRRL PRTWS532GRRS PRTWS532NARXXXL PRTWS532GRRXXXL PRTWS532NAR4XL PRTWS532NARL PRTWS532NARM PRTWS532NARS PRTWS532NARXL PRTWS532NARXXL PRTWS532GRRM PRTWS532RERL PRTWS532RERXL PRTWS532RERS PRTWS532RERXXL PRTWS532RERM PRTWS532RERXXXL
Rain jackets, 3-in-1, Orkney, S532
Werkkleding
This 3-in-1 jacket is made of 100% nylon ottoman, with a breathable PU coating. The detachable fleece jacket is made of 100% anti pill polyester.
  • Pack-away hood
  • Two way zip with double storm flap
  • Two chest pockets with zip fastening, two hip pockets with zip fastening
  • Drawstring hood and waist, elasticated cuffs with Velcro adjustable straps
  • Mobile phone pocket
The waterproof nylon shell is designed to protect workers in all industries against driving rain, wind and cold.
Outside fabric weight: 170 g/m²
Inside fleece jacket weight: 280 g/m²
Certificaten: EN 343 (3-2)