Safety helmet with visor, Endurance, PW55
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PW55RBR PW55RER PW55WHR PW55YER
PRTWPW55RBREA 22.2 EUR
PRTWPW55RBR PRTWPW55RER PRTWPW55WHR PRTWPW55YER
Safety helmet with visor, Endurance, PW55
Hoofdbescherming Veiligheidshelmen
The innovative PW55 has a safety retractable visor attached to the helmet. The shell is made with a durable injection-moulded, high-density ABS material.
  • Equipped with a six-point self-sizing wheel ratchet suspension system
  • Wide nylon straps (2 cm), absorbent crown and brow pad
  • Suitable to be worn in temperatures of −30 °C
  • Compatible with clip on ear protectors: PW42
Size: 520 to 620 mm
Certificaten: EN 397; EN166 1B AS, AF