Trousers, 3/4 length, Originals 283 FAS
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
100291940C44 100291940C46 100291940C48 100291940C50 100291940C52 100291940C54 100291940C56 100291940C58 100291940C60 100291940C62
KWIN100291940C44EA 96.9 EUR
KWIN100291940C44 KWIN100291940C46 KWIN100291940C48 KWIN100291940C50 KWIN100291940C52 KWIN100291940C54 KWIN100291940C56 KWIN100291940C58 KWIN100291940C60 KWIN100291940C62
Trousers, 3/4 length, Originals 283 FAS
Werkkleding
Durable, shorter working trousers made of FAS®, Fristads best twill manufactured from 100% cotton.
  • Rounded knees, Corudra® reinforced knee pockets with opening for height-adjustable knee pads
  • Adjustable leg bottoms
  • Two tuckable, Cordura® reinforced loose-hanging pockets, one with extra pocket, one with three small pockets and tool loops, two front and back pockets with bellows pleat
  • Leg pocket with pocket and tool pocket, folding ruler pocket over side seam
  • Concealed ID-card holder, tool pocket, pen pocket, button and loop for sheath knife, two hammer loops and D-rings
Weight: 375 g/m²