Rain trousers, Le Havre 5100
Leverancier: SIOEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5100/A98/1XL 5100/A98/2XL 5100/A98/3XL 5100/A98/L 5100/A98/M 5100/A98/S
SIOE5100/A98/1XLEA 23.7 EUR
SIOE5100/A98/1XL SIOE5100/A98/2XL SIOE5100/A98/3XL SIOE5100/A98/L SIOE5100/A98/M SIOE5100/A98/S
Rain trousers, Le Havre 5100
Werkkleding
Water- and windproof trousers made of Texoflex fabric: 100% polyester knitting with 100% PVC coating.
  • Tear resistant, extremely strong
  • High frequency welded seams
  • Legs adjustable by press studs
  • Elasticated waist
Weight: 380 g/m²
Certificaten: ENV 343 (3-1)