Nitrification inhibitor for BOD
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
253335 45901 253334 257924
662-0006EA 60.8 EUR
662-0006 HACH45901 HACH253334 HACH257924
Nitrification inhibitor for BOD
Standaarden Standaarden voor milieuanalyse
TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate.