Capillary columns, TracePLOT™ TG-BOND Msieve 5A
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26003-6010 26003-6040 26003-6070 26003-6100 26003-1630
555-8876EA 1240 EUR
555-8876 555-8875 555-8874 555-8873 555-8877
Capillary columns, TracePLOT™ TG-BOND Msieve 5A
Kolommen voor chromatografie
TracePLOT™ TG-BOND Msieve 5A GC columns are designed for separation of Ar/O₂ and other permanent gases.
  • Specially designed coating and deactivation procedures for chromatographic efficiency and the integrity of the coating porous layer
  • Deactivation process yields a sharp peak for CO elution rather than the tailing commonly seen in other columns
  • High retention of molecular sieve permits separation of permanent gases at temperatures above ambient
  • Uniform particles remain adherent to the tubing even following continuous valve-cycling
Order Now
Specificaties
Environmentally Preferable