Capillary columns, TracePLOT™ TG-BOND Q
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26004-6030 26004-6060 26004-6000 26004-6090
555-8871EA 969 EUR
555-8871 555-8870 555-8872 555-8868
Capillary columns, TracePLOT™ TG-BOND Q
Kolommen voor chromatografie
Non-polar columns for oxygenated compounds and solvents.
  • Non-polar 100% divinyl benzene phase
  • Particles incorporated into the walls of the tubing for essentially no particle release