Capillary columns, TracePLOT™ TG-BOND U
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26007-6090 26007-6000 26007-6030
555-8856EA 1110 EUR
555-8856 555-8859 555-8858
Capillary columns, TracePLOT™ TG-BOND U
Kolommen voor chromatografie
TracePLOT™ TG-BOND U GC columns are ideal for the analysis of polar compounds.
  • Polar, divinylbenzene ethylene glycol/dimethylacrylate phase
  • Particles incorporated into the walls of the tubing for essentially no particle release