Microscope objective lens mounts
Leverancier: DIDALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
POD060550 POD060560
DIDAPOD060550EA 91.9 EUR
DIDAPOD060550 DIDAPOD060560
Microscope objective lens mounts
Microscoopobjectieven
The objective holder allows precise positioning of a microscope objective according to the vertical and the horizontal.
  • Both axes 2 mm each
  • Holder is mounted on a 10 mm diameter rod