Pipette tips, HLT, HydroLogix™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3680-11 3681 3950-11 3951-11 3895 3896 3920-11 3800-11 3801-11 3802-11 3880 3881 3890 3980 3682-11
613-2800EA 0 EUR
613-2800 613-2801 613-7435 613-7436 MLBP3895 613-7428 613-7433 MLBP3800-11 MLBP3801-11 613-7431 613-7426 613-7427 MLBP3890 613-2785 MLBP3682-11
Pipette tips, HLT, HydroLogix™
Pipettips
These tips feature a unique bi-fabricated wall that has been designed to produce a secure seal at a fraction of the force required to load standard pipette tips onto the pipettor. Accordingly, the forces required to eject the tips from your pipettor are significantly reduced. Molded from virgin polypropylene, they don't use additives or coatings that may affect sample integrity.