Cleanroom overalls with open legs, 8R013
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1006515002XL 1006515003XL 100651500L 100651500M 100651500S 100651500XL 100651500XS 1006519002XL 1006519003XL 100651900L 100651900M 100651900S 100651900XL 100651900XS
KWIN1006515002XLEA 102.9 EUR
KWIN1006515002XL KWIN1006515003XL KWIN100651500L KWIN100651500M KWIN100651500S KWIN100651500XL KWIN100651500XS KWIN1006519002XL KWIN1006519003XL KWIN100651900L KWIN100651900M KWIN100651900S KWIN100651900XL KWIN100651900XS
Cleanroom overalls with open legs, 8R013
Werkkleding
These overalls are made of XR50 fabric consisting of 97% polyester and 3% conductive fibre. The fabric is ideal for sterile environments.
  • Fabric shows no change in pore size after 50 wash and autoclave cycles
  • Concealed zip, rounded neck with snap fastener
  • Elasticated cuffs, open legs
  • Adjustable elasticated waistband
  • Raglan sleeves
Suitable for use in ISO class 3 cleanrooms.
Weight: 110 g/m²
Order Now
Specificaties
Model Open legs, 8R013, XR50
Environmentally Preferable
Kledingstuk toepassing Cleanroom
Risk area Anti-static
Geslacht
Sluitingstype Met ritssluiting
Gewicht 110 g/m²