Workman cotton shirts, 7388 KC
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1007365402XL 1007365403XL 1007365404XL 100736540L 100736540M 100736540S 100736540XL
KWIN1007365402XLEA 59.9 EUR
KWIN1007365402XL KWIN1007365403XL KWIN1007365404XL KWIN100736540L KWIN100736540M KWIN100736540S KWIN100736540XL
Workman cotton shirts, 7388 KC
Werkkleding
Durable shirts made of 100% combed cotton.
  • Drawstring at neck
  • Inside pocket
  • Sleeve pocket
  • Two side pockets
Weight: 350 g/m²