HPLC columns, Hypersil™ Green PAH
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31103-103030 31105-254630 31105-154630 31105-104630 31105-014001 31103-154630 31103-013001 31103-153030 31103-152130 31105-253030 31103-104630 31103-104030 31103-102130 31103-053030 31103-014001 31103-012101
555-5292EA 1140 EUR
555-5292 555-5328 555-5332 555-5336 555-5283 555-5286 555-5300 555-5288 555-5289 555-5330 555-5290 555-5291 555-5293 555-5296 555-5299 555-5301
HPLC columns, Hypersil™ Green PAH
Kolommen voor chromatografie
The Hypersil™ green PAH columns with alkyl bonded silica and high carbon loading are suitable for the analysis of polyaromatic hydrocarbons (PAHs).
  • Optimised for EPA method 610
  • Rapid analysis of 16 PAHs using short, fast columns