HPLC columns, Hypersil™ APS-2
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30703-154630 30703-054630 30705-154630 30705-014001 30705-254630 30705-254030 30703-012101 30703-152130 30703-102130 30703-014001 30705-103030
555-5913EA 887 EUR
555-5913 555-5923 555-6735 555-5901 555-6702 555-6703 555-5988 555-5916 555-5921 555-5986 555-6742
HPLC columns, Hypersil™ APS-2
Kolommen voor chromatografie
Hypersil™ APS-2 is an amino propyl phase column and is suitable for analysis of sugars.