HPLC columns, Hypersil™ MOS and MOS-2 C8
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30205-254030 30305-254630 30305-154630 30205-254630 30205-204630 30205-154630 30205-104630 30205-014001 30305-014001 30205-253030 30205-154030 30205-054630
555-6251EA 840 EUR
555-6251 555-5974 555-5980 555-6250 555-6254 555-6256 555-6262 555-6329 555-6276 555-6252 555-6257 555-6324
HPLC columns, Hypersil™ MOS and MOS-2 C8
Kolommen voor chromatografie
Hypersil™ MOS and MOS-2 columns have a monolayer coverage of C8 alkyl chain chemically bonded onto the silica surface for a reproducible and efficient stationary phase. The MOS-2 is endcapped. Both C8 phases exhibit similar selectivity to ODS, but are less retentive. This can often provide reduced time for analyses, particularly for strongly hydrophobic analytes.
  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Dependable and affordable