Cleanroom coats, short
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1006324102XL 100632410L 100632410M 100632410S 100632410XL 100632410XS
KWIN1006324102XLEA 78.5 EUR
KWIN1006324102XL KWIN100632410L KWIN100632410M KWIN100632410S KWIN100632410XL KWIN100632410XS
Cleanroom coats, short
Werkkleding
These coats are made of XA32 fabric consisting of 99% polyester and 1% conductive fibres. The fibres are continuous filament fibres, which means that the garments do not emit particles or fibres. The carbon filaments produce good static discharge, thus preventing static charging and minimising triboelectric attraction of particles and fibres. This makes the garments very suitable for activities such as vehicle spraying.
  • Concealed zip
  • Crew neck with snap fastener
  • Elasticated cuffs
  • Raglan sleeves
Weight: 125 g/m²