Cyclopentylboronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

C2442-1G C2442-5G
TCIAC2442-1GEA 24.2 EUR
TCIAC2442-1G TCIAC2442-5G
Cyclopentylboronic acid (contains varying amounts of Anhydride)
Cyclopentylboronic acid
Formule: C₅H₁₁BO₂
MDL Nummer: MFCD01074541
CAS nummer: 63076-51-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ce1215e36f8ba04c1de2df18380cb5a6fe1433b&vid=0ce1215e36f8ba04c1de2df18380cb5a6fe1433b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 119.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR