N-Fmoc-DL-norleucine

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4025138.0005 4025138.0025
BACE4025138.0005EA 0 EUR
BACE4025138.0005 BACE4025138.0025
N-Fmoc-DL-norleucine
N-Fmoc-DL-norleucine
Formule: C₂₁H₂₃NO₄
MDL Nummer: MFCD00237391
CAS nummer: 144701-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR