1-Boc-4-piperidineacetic acid ≥97.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

B3703-1G B3703-5G
TCIAB3703-1GEA 19.8 EUR
TCIAB3703-1G TCIAB3703-5G
1-Boc-4-piperidineacetic acid ≥97.0% (by GC, titration analysis)
1-Boc-4-piperidineacetic acid
Formule: C₁₂H₂₁NO₄
MDL Nummer: MFCD00800239
CAS nummer: 157688-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=145f35c38efb23bbf2d6c2ac6d6cc78ddcd20f04&vid=145f35c38efb23bbf2d6c2ac6d6cc78ddcd20f04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 97.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR