(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (3S)-1,2,3,4-Tetrahydro-3-isoquinolinecarbocylic acid, (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid

4016025.0005 4016025.0025
BACE4016025.0005EA 0 EUR
BACE4016025.0005 BACE4016025.0025
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Formule: C₁₀H₁₁NO₂
MW: 177.20 g/mol
MDL Nummer: MFCD00144533
CAS nummer: 74163-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR