(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

T1515-25G T1515-5G
TCIAT1515-25GEA 51.7 EUR
TCIAT1515-25G TCIAT1515-5G
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid ≥98.0% (by titrimetric analysis)
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Formule: C₁₀H₁₁NO₂
MDL Nummer: MFCD00144533
CAS nummer: 74163-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df87b85376f80a57c99b20bbcd3c20e7921848e4&vid=df87b85376f80a57c99b20bbcd3c20e7921848e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -163 to -169 deg (C=1, 1mol/L NaOH sol.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR