3,5-Dimethylisoxazole-4-boronic acid pinacol ester ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole

D3772-1G D3772-5G
TCIAD3772-1GEA 39.6 EUR
TCIAD3772-1G TCIAD3772-5G
3,5-Dimethylisoxazole-4-boronic acid pinacol ester ≥98.0% (by GC)
3,5-Dimethylisoxazole-4-boronic acid pinacol ester
Formule: C₁₁H₁₈BNO₃
MW: 223.08 g/mol
MDL Nummer: MFCD05863910
CAS nummer: 832114-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8ab120965ff0a6f8f636b3ab05e70f6a92efce4&vid=c8ab120965ff0a6f8f636b3ab05e70f6a92efce4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 101.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR