5-(4-Bromophenyl)isoxazole
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CC12910DA
CC12910EA
MAYBCC12910DAEA
0
EUR
MAYBCC12910DA
MAYBCC12910EA
5-(4-Bromophenyl)isoxazole
5-(4-Bromophenyl)isoxazole
Formule:
C₉H₆BrNO MW: 224.06 g/mol |
MDL Nummer:
MFCD02183532 CAS nummer: 7064-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...