N-(2-Bromoethyl)-2,4-dichlorobenzenesulphonamide
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB09486.250 BTB09486.1
MAYBBTB09486.250EA 0 EUR
MAYBBTB09486.250 MAYBBTB09486.1
N-(2-Bromoethyl)-2,4-dichlorobenzenesulphonamide
N-(2-Bromoethyl)-2,4-dichlorobenzenesulphonamide
Formule: C₈H₈BrCl₂NO₂S
CAS nummer: 680579-43-5