N-{[3,5-Bis(trifluoromethyl)phenyl]sulphonyl}-N-(2,6-dibromo-4-methylphenyl)-3,5-bis(trifluoromethyl)benzenesulphonamide
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SEW05306.250 SEW05306.1
MAYBSEW05306.250EA 0 EUR
MAYBSEW05306.250 MAYBSEW05306.1
N-{[3,5-Bis(trifluoromethyl)phenyl]sulphonyl}-N-(2,6-dibromo-4-methylphenyl)-3,5-bis(trifluoromethyl)benzenesulphonamide
N-{[3,5-Bis(trifluoromethyl)phenyl]sulphonyl}-N-(2,6-dibromo-4-methylphenyl)-3,5-bis(trifluoromethyl)benzenesulphonamide
Formule: C₂₃H₁₁Br₂F₁₂NO₄S₂
MDL Nummer: MFCD00662513
CAS nummer: 259269-91-5