2-[(3-Fluorobenzyl)oxy]-5-nitrobenzaldehyde
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BTB05372.1 BTB05372.250
MAYBBTB05372.1EA 0 EUR
MAYBBTB05372.1 MAYBBTB05372.250
2-[(3-Fluorobenzyl)oxy]-5-nitrobenzaldehyde
2-[(3-Fluorobenzyl)oxy]-5-nitrobenzaldehyde
Formule: C₁₄H₁₀FNO₄
MDL Nummer: MFCD00117030
CAS nummer: 219539-49-8